Få mere at vide
2-Chloro-2-methylpropane-d9, TRC
Artikelnummer. 30368650
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30368650 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30368650 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3696821G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Chloro-2-methylpropane-d9
CAS 918-20-7
Molekylær formel C4 D9 Cl
Mængde 1 g
Synonym 1,1-Dimethylethyl-d9 Chloride, 1-Chloro-1,1-dimethylethane-d9, 2-Chloro-2-methylpropane-d9, 2-Chloroisobutane-d9, 2-Methyl-2-chloropropane-d9, 2-Methyl-2-propyl Chloride-d9, Chlorotrimethylmethane-d9, NSC 6527-d9, Trimethylchloromethane-d9, tert-Butyl-d9 Chloride
InChI formel InChI=1S/C4H9Cl/c1-4(2,3)5/h1-3H3/i1D3,2D3,3D3
SMIL [2H]C([2H])([2H])C(Cl)(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 2-chloro-1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propane
Molekylvægt (g/mol) 101.62
Formel vægt 101.0958