Learn More
2-Chloro-2',4'-difluoroacetophenone, Mikromol™
Product Code. 30459082
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30459082 4 x 25 mg -
30051650 100 mg 100mg
30050727 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 30459082 Supplier Mikromol Supplier No. MM0235.0400254AMP
 Restricted Item
Forventet forsendelse: 30-11-2026
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Chloro-2',4'-difluoroacetophenone, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Chloro-2',4'-difluoroacetophenone
CAS 51336-94-8
Analyte or Component Names 2-Chloro-2',4'-difluoroacetophenone
Shipping Condition Room Temperature
Molecular Formula C8 H5 Cl F2 O
Certifications/Compliance ISO 17025
Synonym 2-Chloro-2',4'-difluoroacetophenone
InChI Formula InChI=1S/C8H5ClF2O/c9-4-8(12)6-2-1-5(10)3-7(6)11/h1-3H,4H2
Solution Type Neat
SMILES Fc1ccc(C(=O)CCl)c(F)c1
IUPAC Name 2-chloro-1-(2,4-difluorophenyl)ethanone
Molecular Weight (g/mol) 190.57
Quantity 4 x 25 mg
Formula Weight 189.9997 g/mol