missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1-methylpyridinium iodide, 97%, Thermo Scientific Chemicals

Catalog Number p-7020757
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
14338-32-0
C6H7ClIN
255.48
MFCD00011984
ABFPKTQEQNICFT-UHFFFAOYSA-M
2-chloro-1-methylpyridinium iodide, 2-chloro-1-methylpyridin-1-ium iodide, pyridinium, 2-chloro-1-methyl-, iodide, unii-rlq88nyz68, rlq88nyz68, 2-chloro-1-methylpyridiniumiodide, 2-chloro-1-methyl-pyridinium iodide, pubchem16659, acmc-1brmd, ksc494o1l
167069
2-chloro-1-methylpyridin-1-ium;iodide
[I-].C[N+]1=CC=CC=C1Cl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
14338-32-0
C6H7ClIN
255.48
MFCD00011984
ABFPKTQEQNICFT-UHFFFAOYSA-M
2-chloro-1-methylpyridinium iodide, 2-chloro-1-methylpyridin-1-ium iodide, pyridinium, 2-chloro-1-methyl-, iodide, unii-rlq88nyz68, rlq88nyz68, 2-chloro-1-methylpyridiniumiodide, 2-chloro-1-methyl-pyridinium iodide, pubchem16659, acmc-1brmd, ksc494o1l
167069
2-chloro-1-methylpyridin-1-ium;iodide
[I-].C[N+]1=CC=CC=C1Cl

2-Chloro-1-methylpyridinium iodide is a reagent in a wide variety of dehydrative coupling reactions. It is also used to form carboxylate esters from acids and alcohols, carboxamides from acids and amines, lactones from -hydroxy acids and carbodiimides from N,N-disubstituted thioureas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-1-methylpyridinium iodide is a reagent in a wide variety of dehydrative coupling reactions. It is also used to form carboxylate esters from acids and alcohols, carboxamides from acids and amines, lactones from -hydroxy acids and carbodiimides from N,N-disubstituted thioureas.

Solubility
Soluble in Methanol (50 mg/ml). Insoluble in water.

Notes
Store at 4°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point ∼200°C (decomposition)
Quantity 25 g
Beilstein 3572320
Sensitivity Light and moisture sensitive
Merck Index 14,6301
Solubility Information Soluble in Methanol (50mg/ml). Insoluble in water.
Formula Weight 255.49
Percent Purity 97%
Chemical Name or Material 2-Chloro-1-methylpyridinium iodide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'