missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Carboxythiophene-3-boronic Acid, TRC

Product Code. 30340812
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30340812 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30340812 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCB41593850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2-Carboxythiophene-3-boronic Acid
CAS 5503-74-2
Molekylær formel C5H5BO4S
Mængde 50 mg
Synonym 3-Borono-2-thiophenecarboxylic acid
InChI formel InChI=1S/C5H5BO4S/c7-5(8)4-3(6(9)10)1-2-11-4/h1-2,9-10H,(H,7,8)
SMIL OB(O)c1ccsc1C(=O)O
IUPAC navn 3-boronothiophene-2-carboxylic acid
Molekylvægt (g/mol) 171.97
Formel vægt 172.0002
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.