Få mere at vide
(2-Carboxyethyl)dimethylsulfonium Chloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Dimethylsulfoniopropionate hydrochloride |
| CAS | 4337-33-1 |
| Molekylær formel | C5 H11 O2 S . Cl |
| Mængde | 100 g |
| Synonym | DMSP hydrochloride, beta-DMSP hydrochloride, (2-Carboxyethyl)dimethyl sulfonium chloride, Dimethyl-beta-propiothetin chloride, Dimethyl-beta-propiothetin hydrochloride, Dimethyl-3-propiothetin hydrochloride, beta-Dimethylsulfoniopropionate hydrochloride, 3-Dimethylsulfoniopropionate hydrochloride, S,S-Dimethyl-beta-propiothetin hydrochloride |
| InChI formel | InChI=1S/C5H10O2S.ClH/c1-8(2)4-3-5(6)7;/h3-4H2,1-2H3;1H |
| SMIL | [Cl-].C[S+](C)CCC(=O)O |
| IUPAC navn | 2-carboxyethyl(dimethyl)sulfanium;chloride |
| Molekylvægt (g/mol) | 170.66 |
| Formel vægt | 170.0168 |