Få mere at vide
(2-Carboxyethyl)dimethyl-d6-sulfonium Chloride, TRC
Artikelnummer. 30363611
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30363611 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30363611 Leverandør Toronto Research Chemicals Leverandørnr. TRCC1780625MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2-Carboxyethyl)dimethyl-d6-sulfonium Chloride
CAS 1246274-70-3
Molekylær formel C5 D6 H5 O2 S . Cl
Mængde 5 mg
Synonym Sulfonium, (2-carboxyethyl)di(methyl-d3)-, chloride (1:1) (ACI), (2-Carboxyethyl)dimethyl-d6-sulfonium Chloride, (2-Carboxyethyl)dimethylsulfonium-d6 chloride, (2-Carboxyethyl)dimethylsulfonium-D6 (S,S-dimethyl-D6) Chloride, (2-Carboxyethyl)di(methyl-d3)sulfonium chloride, 3-Di(methyl-d3)sulphoniopropionate hydrochloride, Dimethyl-β-propiothetin-d6 chloride, Dimethyl-β-propiothetin-d6 hydrochloride, Dimethyl-d6-propiothetin Hydrochloride
InChI formel InChI=1S/C5H10O2S.ClH/c1-8(2)4-3-5(6)7;/h3-4H2,1-2H3;1H/i1D3,2D3;
SMIL [Cl-].[2H]C([2H])([2H])[S+](CCC(=O)O)C([2H])([2H])[2H]
IUPAC navn 2-carboxyethyl-bis(trideuteriomethyl)sulfanium;chloride
Molekylvægt (g/mol) 176.695
Formel vægt 176.054