Få mere at vide
(2-Carboxyethyl)dimethyl-d6-sulfonium Chloride, CDN
Artikelnummer. 30312295
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30312295 0.1 g 0.1g
30311333 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312295 Leverandør CDN Leverandørnr. CDND78780.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2-Carboxyethyl)dimethyl-d6-sulfonium Chloride
CAS 1246274-70-3
Molekylær formel C5 D6 H5 O2 S . Cl
Mængde 0.1 g
Synonym Sulfonium, (2-carboxyethyl)di(methyl-d3)-, chloride (1:1) (ACI), (2-Carboxyethyl)dimethyl-d6-sulfonium Chloride, (2-Carboxyethyl)dimethylsulfonium-d6 chloride, (2-Carboxyethyl)dimethylsulfonium-D6 (S,S-dimethyl-D6) Chloride, (2-Carboxyethyl)di(methyl-d3)sulfonium chloride, 3-Di(methyl-d3)sulphoniopropionate hydrochloride, Dimethyl-β-propiothetin-d6 chloride, Dimethyl-β-propiothetin-d6 hydrochloride, Dimethyl-d6-propiothetin Hydrochloride
InChI formel InChI=1 S/C5H10O2S.ClH/c1-8(2)4-3-5(6)7;/h3-4H2,1-2H3;1 H/i1D3,2D3;
SMIL [Cl-].[2 H]C([2 H])([2 H])[S+](CCC(=O)O)C([2 H])([2 H])[2 H]
IUPAC navn 2-carboxyethyl-bis(trideuteriomethyl)sulfanium;chloride
Molekylvægt (g/mol) 176.695
Formel vægt 176.054 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity