Få mere at vide
2-Butyl-d3-4-chloro-5-benzyloxymethyl-1H-imidazole, TRC
Artikelnummer. 30333847
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30333847 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30333847 Leverandør Toronto Research Chemicals Leverandørnr. TRCB69060725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Butyl-d3-4-chloro-5-benzyloxymethyl-1H-imidazole
CAS 1189919-69-4
Molekylær formel C15H16D3ClN2O
Mængde 25 mg
Synonym 2-Butyl-d3-5-chloro-4-[(phenylmethoxy)methyl]-1H-imidazole, 2-Butyl-d3-4-chloro-5-[(phenylmethoxy)methyl]-1H-imidazole
InChI formel InChI=1S/C15H19ClN2O/c1-2-3-9-14-17-13(15(16)18-14)11-19-10-12-7-5-4-6-8-12/h4-8H,2-3,9-11H2,1H3,(H,17,18)/i1D3
SMIL [2H]C([2H])([2H])CCCc1nc(COCc2ccccc2)c(Cl)[nH]1
IUPAC navn 5-chloro-4-(phenylmethoxymethyl)-2-(4,4,4-trideuteriobutyl)-1H-imidazole
Molekylvægt (g/mol) 281.8
Formel vægt 281.1374