Få mere at vide
2-Butyl-d3-4-chloro-1H-imidazole-5-carboxaldehyde, TRC
Artikelnummer. 30340818
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30340818 2.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30340818 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6906322.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Butyl-d3-4-chloro-1H-imidazole-5-carboxaldehyde
CAS 1189927-81-8
Molekylær formel C8H8D3ClN2O
Mængde 2.5 mg
Synonym 2-Butyl-d3-5-chloro-1H-imidazole-4-carboxaldehyde, 2-Butyl-d3-4-chloro-5-formylimidazole, 2-Butyl-d3-4-chloroimidazole-5-carboxaldehyde, 2-Butyl-d3-5-formyl-4-chloroimidazole
InChI formel InChI=1S/C8H11ClN2O/c1-2-3-4-7-10-6(5-12)8(9)11-7/h5H,2-4H2,1H3,(H,10,11)/i1D3
SMIL [2H]C([2H])([2H])CCCc1nc(C=O)c(Cl)[nH]1
IUPAC navn 5-chloro-2-(4,4,4-trideuteriobutyl)-1H-imidazole-4-carbaldehyde
Molekylvægt (g/mol) 189.66
Formel vægt 189.0748