Få mere at vide
2-Butyl-4-chloro-1H-imidazole-5-carbaldehyde, Mikromol™
Artikelnummer. 30049693
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049693 100 mg
30051754 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30049693 Leverandør Mikromol Leverandørnr. MM0168.08
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Butyl-4-chloro-1H-imidazole-5-carbaldehyde
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Butyl-4-chloro-1H-imidazole-5-carbaldehyde
CAS 83857-96-9
Analyt- eller komponentnavne 2-Butyl-4-chloro-1H-imidazole-5-carbaldehyde
Forsendelsestilstand Room Temperature
Molekylær formel C8 H11 Cl N2 O
Certificeringer/compliance ISO 17025
Synonym Losartan Potassium Imp. D (EP), Losartan Imp. D (EP), Losartan USP Related Compound A, Losartan USP RC A, 2-Butyl-4-chloro-1H-imidazole-5-carbaldehyde, Losartan Related Compound A, Losartan Potassium Impurity D, Losartan Impurity D
InChI formel InChI=1S/C8H11ClN2O/c1-2-3-4-7-10-6(5-12)8(9)11-7/h5H,2-4H2,1H3,(H,10,11)
SMIL CCCCc1nc(Cl)c(C=O)[nH]1
IUPAC navn 2-butyl-4-chloro-1H-imidazole-5-carbaldehyde
Molekylvægt (g/mol) 186.64
Mængde 100 mg
Formel vægt 186.056 g/mol