Få mere at vide
[2-Butyl-4-chloro-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl]biphenyl-4-yl]methyl]-1H-imidazol-5-yl]methanol, Mikromol™
Artikelnummer. 30459766
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30459766 4 x 25 mg
30050297 100 mg
30049374 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30459766 Leverandør Mikromol Leverandørnr. MM0168.1600254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
[2-Butyl-4-chloro-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl]biphenyl-4-yl]methyl]-1H-imidazol-5-yl]methanol, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2-Butyl-4-chloro-1-[(2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl]biphenyl-4-yl)methyl]-1H-imidazol-5-yl)methanol
CAS 133909-99-6
Analyt- eller komponentnavne (2-Butyl-4-chloro-1-[(2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl]biphenyl-4-yl)methyl]-1H-imidazol-5-yl)methanol
Forsendelsestilstand Room Temperature
Molekylær formel C41 H37 Cl N6 O
Certificeringer/compliance ISO 17025
Synonym Losartan Potassium Imp. H (EP), Losartan Imp. H (EP), [2-Butyl-4-chloro-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl]biphenyl-4-yl]methyl]-1H-imidazol-5-yl]methanol, Losartan Potassium Impurity H, Losartan Impurity H
InChI formel InChI=1S/C41H37ClN6O/c1-2-3-23-38-43-39(42)37(29-49)47(38)28-30-24-26-31(27-25-30)35-21-13-14-22-36(35)40-44-46-48(45-40)41(32-15-7-4-8-16-32,33-17-9-5-10-18-33)34-19-11-6-12-20-34/h4-22,24-27,49H,2-3,23,28-29H2,1H3
Løsningstype Neat
SMIL CCCCc1nc(Cl)c(CO)n1Cc2ccc(cc2)c3ccccc3c4nnn(n4)C(c5ccccc5)(c6ccccc6)c7ccccc7
IUPAC navn [2-butyl-5-chloro-3-[[4-[2-(2-trityltetrazol-5-yl)phenyl]phenyl]methyl]imidazol-4-yl]methanol
Molekylvægt (g/mol) 665.23
Mængde 4 x 25 mg
Formel vægt 664.2717 g/mol