Få mere at vide
2-Butanone-d8, CDN
Artikelnummer. 30321543
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321543 0.05 g
30317763 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321543 Leverandør CDN Leverandørnr. CDND43310.05G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Butanone-D8
CAS 350820-09-6
Molekylær formel C4 D8 O
Mængde 0.05 g
Synonym 2-Butanone-1,1,1,3,3,4,4,4-d8 (9 CI), 2-Butanone-d8, 2-Butanone-D8, Methyl-d3 ethyl-d5 ketone
InChI formel InChI=1 S/C4H8O/c1-3-4(2)5/h3H2,1-2H3/i1D3,2D3,3D2
SMIL [2 H]C([2 H])([2 H])C(=O)C([2 H])([2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1,1,1,3,3,4,4,4-octadeuteriobutan-2-one
Molekylvægt (g/mol) 80.155
Formel vægt 80.1077 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity