Få mere at vide
2-Butanone-4,4,4-d3, CDN
Artikelnummer. 30312606
Skift visning
Mængde:
0.1 g
Pakningsstørrelse:
0.1g
Artikelnummer. Mængde unitSize
30312606 0.1 g 0.1g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312606 Leverandør CDN Leverandørnr. CDND24020.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Butanone 4,4,4-d3
CAS 53389-26-7
Molekylær formel C4 D3 H5 O
Mængde 0.1 g
Synonym Butanone D3 (4,4,4 D3), 2-Butanone-4,4,4-d3, 2-Butanone D3 (4,4,4-D3)
InChI formel InChI=1 S/C4H8O/c1-3-4(2)5/h3H2,1-2H3/i1D3
SMIL [2 H]C([2 H])([2 H])CC(=O)C
IUPAC navn 4,4,4-trideuteriobutan-2-one
Molekylvægt (g/mol) 75.1242
Formel vægt 75.0763 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity