Få mere at vide
2-Butanone-1,1,1,3,3-d5, CDN
Artikelnummer. 30320101
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30320101 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320101 Leverandør CDN Leverandørnr. CDND02931G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Butanone 1,1,1,3,3-d5
CAS 24313-50-6
Molekylær formel C4 2H5 H3 O
Mængde 1 g
Synonym 2-Butanone-1,1,1,3,3-d5 (6 CI,8 CI,9 CI), 2-Butanone-d5
InChI formel InChI=1 S/C4H8O/c1-3-4(2)5/h3H2,1-2H3/i2D3,3D2
SMIL [2 H]C([2 H])([2 H])C(=O)C([2 H])([2 H])C
IUPAC navn 1,1,1,3,3-pentadeuteriobutan-2-one
Molekylvægt (g/mol) 77.14
Formel vægt 77.0889 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity