Få mere at vide
2-Butanol-d3, TRC
Artikelnummer. 30338854
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30338854 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30338854 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68989725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Butanol-d3
CAS 53716-61-3
Molekylær formel C4 D3 H7 O
Mængde 25 mg
Synonym sec-Butyl-2',2',2'-d3 Alcohol, 2-Butan-1,1,1-d3-ol, 2-Butan-1,1,1-d3-ol (9CI), sec-Butyl-2',2',2'-d3 alcohol (6CI), 1,1,1-Trideuteriobutan-2-ol, (±)-sec-Butyl-1,1,1-d3 alcohol, 2-Butanol-d3, Butan-2-ol-1,1,1-D3
InChI formel InChI=1S/C4H10O/c1-3-4(2)5/h4-5H,3H2,1-2H3/i2D3
SMIL [2H]C([2H])([2H])C(O)CC
IUPAC navn 1,1,1-trideuteriobutan-2-ol
Molekylvægt (g/mol) 77.14
Formel vægt 77.092