Få mere at vide
2-Butan-3,3,4,4,4-d5-ol, TRC
Artikelnummer. 30329791
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30329791 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30329791 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68989650MG
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Butan-3,3,4,4,4-d5-ol
CAS 75749-92-7
Molekylær formel C4 D5 H5 O
Mængde 50 mg
Synonym 2-Butan-3,3,4,4,4-d5-ol (9CI), sec-Butyl-d5 alcohol, 3,3,4,4,4-Pentadeuteriobutan-2-ol, (±)-sec-Butyl-3,3,4,4,4-d5 alcohol, 2-Butanol-d5, Butan-2-ol-3,3,4,4,4-D5
InChI formel InChI=1S/C4H10O/c1-3-4(2)5/h4-5H,3H2,1-2H3/i1D3,3D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C(C)O
IUPAC navn 3,3,4,4,4-pentadeuteriobutan-2-ol
Molekylvægt (g/mol) 79.1524
Formel vægt 79.1045