Få mere at vide
2-Bromotoluene-d7, CDN
Artikelnummer. 30317625
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30317625 0.25 g
30310072 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317625 Leverandør CDN Leverandørnr. CDND64040.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Bromotoluene-D7
CAS 1185306-95-9
Molekylær formel C7 D7 Br
Mængde 0.25 g
Synonym 5-Bromo-6-(methyl-d3)benzene-1,2,3,4-d4, 2-Bromotoluene-d7, 2-Bromotoluene-D7, 2-Bromotoluene-α,α,α,3,4,5,6-d7
InChI formel InChI=1 S/C7H7Br/c1-6-4-2-3-5-7(6)8/h2-5 H,1H3/i1D3,2 D,3 D,4 D,5 D
SMIL [2 H]c1c([2 H])c([2 H])c(c(Br)c1[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1-bromo-2,3,4,5-tetradeuterio-6-(trideuteriomethyl)benzene
Molekylvægt (g/mol) 178.0776
Formel vægt 177.0171 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity