missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothiophene-3-carbonitrile, 97%, Thermo Scientific™

Catalog Number p-3522452
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
56182-43-5
C5H2BrNS
188.042
MFCD09064981
GHMDOVNPFWGFSZ-UHFFFAOYSA-N
2-bromo-3-cyanothiophene, 2-bromo-3-cyanothiophen, 2-bromo-3-thiophenecarbonitrile, 2-bromo-thiophene-3-carbonitrile, 3-thiophenecarbonitrile,2-bromo, 2-bromanylthiophene-3-carbonitrile, 3-thiophenecarbonitrile, 2-bromo
12079026
2-bromothiophene-3-carbonitrile
C1=CSC(=C1C#N)Br
This item is not returnable. View return policy
56182-43-5
C5H2BrNS
188.042
MFCD09064981
GHMDOVNPFWGFSZ-UHFFFAOYSA-N
2-bromo-3-cyanothiophene, 2-bromo-3-cyanothiophen, 2-bromo-3-thiophenecarbonitrile, 2-bromo-thiophene-3-carbonitrile, 3-thiophenecarbonitrile,2-bromo, 2-bromanylthiophene-3-carbonitrile, 3-thiophenecarbonitrile, 2-bromo
12079026
2-bromothiophene-3-carbonitrile
C1=CSC(=C1C#N)Br
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 188.05
Percent Purity 97%
Chemical Name or Material 2-Bromothiophene-3-carbonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'