Learn More
2-Bromoquinoline, 98%, Thermo Scientific Chemicals
Catalog Number p-7077688
Click to view available options
Quantity:
1 g
5 g
25 g
2005-43-8
C9H6BrN
208.06
MFCD00234480
QKJAZPHKNWSXDF-UHFFFAOYSA-N
quinoline, 2-bromo, bromoquinoline, pubchem21827, acmc-1cfcf, 2-bromoquinoline, ksc201o8j, 2-bromoquinoline 1g, abbypharma ap-12-10890
2762756
2-bromoquinoline
BrC1=CC=C2C=CC=CC2=N1
2005-43-8
C9H6BrN
208.06
MFCD00234480
QKJAZPHKNWSXDF-UHFFFAOYSA-N
quinoline, 2-bromo, bromoquinoline, pubchem21827, acmc-1cfcf, 2-bromoquinoline, ksc201o8j, 2-bromoquinoline 1g, abbypharma ap-12-10890
2762756
2-bromoquinoline
BrC1=CC=C2C=CC=CC2=N1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 49.0°C to 51.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 208.06
Percent Purity 98%
Chemical Name or Material 2-Bromoquinoline
compliance-icons
Product Identifier
  • 2-Bromoquinoline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.