Få mere at vide
2-Bromopyridine N-Oxide Hydrochloride, TRC
Artikelnummer. 30347606
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30347606 500 mg
1 options
Artikelnummer. 30347606 Leverandør Toronto Research Chemicals Leverandørnr. TRCB698835500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Bromopyridine N-Oxide Hydrochloride
CAS 80866-91-7
Molekylær formel C5H4BrNO . (HCl)
Mængde 500 mg
Synonym Pyridine, 2-bromo-, 1-oxide, hydrochloride (1:1) (ACI), Pyridine, 2-bromo-, 1-oxide, compd. with hydrochloric acid (7CI), Pyridine, 2-bromo-, 1-oxide, hydrochloride (6CI, 9CI), 2-Bromopyridine 1-oxide hydrochloride, 2-Bromopyridine N-oxide hydrochloride
InChI formel InChI=1S/C5H4BrNO.ClH/c6-5-3-1-2-4-7(5)8;/h1-4H;1H
SMIL Cl.[O-][n+]1ccccc1Br
IUPAC navn 2-bromo-1-oxidopyridin-1-ium;hydrochloride
Molekylvægt (g/mol) 210.45
Formel vægt 208.9243