Få mere at vide
2-Bromopyridine-d4, TRC
Artikelnummer. 30342265
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30342265 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30342265 Leverandør Toronto Research Chemicals Leverandørnr. TRCB686728250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Bromopyridine-d4
CAS 70766-71-1
Molekylær formel C5 2H4 Br N
Mængde 250 mg
Synonym Pyridine-2,3,4,5-d4, 6-bromo- (9CI, ACI), 6-Bromopyridine-2,3,4,5-d4 (ACI), 2-Bromo-3,4,5,6-tetradeuteriopyridine, 2-Bromopyridine-D4, 2-Bromopyridine-d4
InChI formel InChI=1S/C5H4BrN/c6-5-3-1-2-4-7-5/h1-4H/i1D,2D,3D,4D
SMIL [2H]c1nc(Br)c([2H])c([2H])c1[2H]
IUPAC navn 2-bromo-3,4,5,6-tetradeuteriopyridine
Molekylvægt (g/mol) 162.02
Formel vægt 160.9778