Få mere at vide
(±)-2-Bromopropionic-2,3,3,3-d4 Acid, CDN
Artikelnummer. 30322361
Skift visning
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30322361 1 g
30318121 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322361 Leverandør CDN Leverandørnr. CDND55981G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Bromopropanoic Acid-D4
CAS 60153-93-7
Molekylær formel C3 D4 H Br O2
Mængde 1 g
Synonym Propanoic-2,3,3,3-d4 acid, 2-bromo- (9 CI), 2-Bromopropanoic-2,3,3,3-d4 acid, 2-Bromopropanoic acid-2,3,3,3-d4, 2-Bromopropanoic Acid-D4, (±)-2-Bromopropionic-2,3,3,3-d4 Acid
InChI formel InChI=1 S/C3H5BrO2/c1-2(4)3(5)6/h2H,1H3,(H,5,6)/i1D3,2 D
SMIL [2 H]C([2 H])([2 H])C([2 H])(Br)C(=O)O
IUPAC navn 2-bromo-2,3,3,3-tetradeuteriopropanoic acid
Molekylvægt (g/mol) 156.9993
Formel vægt 155.9724 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity