Få mere at vide
2-Bromopropane-d7, TRC
Artikelnummer. 30346553
Skift visning
Mængde:
5 g
Artikelnummer. Mængde
30346553 5 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30346553 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6866275G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Bromopropane-d7
CAS 39091-63-9
Renhedsgrad noter GC
Molekylær formel C3 2H7 Br
Mængde 5 g
Synonym Propane-1,1,1,2,3,3,3-d7, 2-bromo- (9CI), 2-Bromo-1,1,1,2,3,3,3-heptadeuteriopropane, 2-Bromopropane-d7, Isopropyl-d7 bromide, Perdeuterioisopropyl bromide, 2-Bromopropane-D7, 2-Bromopropane-1,1,1,2,3,3,3-d7
InChI formel InChI=1S/C3H7Br/c1-3(2)4/h3H,1-2H3/i1D3,2D3,3D
SMIL [2H]C([2H])([2H])C([2H])(Br)C([2H])([2H])[2H]
IUPAC navn 2-bromo-1,1,1,2,3,3,3-heptadeuteriopropane
Molekylvægt (g/mol) 130.03
Formel vægt 129.0171
Procent renhed >95