Få mere at vide
2-Bromopropane-d7, CDN
Artikelnummer. 30326686
Skift visning
Mængde:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30326686 10 g
30311893 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326686 Leverandør CDN Leverandørnr. CDND075110G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Bromopropane-d7
CAS 39091-63-9
Molekylær formel C3 2H7 Br
Mængde 10 g
InChI formel InChI=1 S/C3H7Br/c1-3(2)4/h3H,1-2H3/i1D3,2D3,3 D
SMIL [2 H]C([2 H])([2 H])C([2 H])(Br)C([2 H])([2 H])[2 H]
IUPAC navn 2-bromo-1,1,1,2,3,3,3-heptadeuteriopropane
Molekylvægt (g/mol) 130.03
Formel vægt 129.0171 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity