Få mere at vide
2-Bromopropane-1,1,1,3,3,3-d6, CDN
Artikelnummer. 30319582
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319582 1 g
30320586 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319582 Leverandør CDN Leverandørnr. CDND07931G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Bromopropane-d6
CAS 52809-76-4
Molekylær formel C3 D6 H Br
Mængde 1 g
Synonym 2-Bromopropane-1,1,1,3,3,3-d6, 2-Bromo-1,1,1,3,3,3-hexadeuteropropane
InChI formel InChI=1 S/C3H7Br/c1-3(2)4/h3H,1-2H3/i1D3,2D3
SMIL [2 H]C([2 H])([2 H])C(Br)C([2 H])([2 H])[2 H]
IUPAC navn 2-bromo-1,1,1,3,3,3-hexadeuteriopropane
Molekylvægt (g/mol) 129.03
Formel vægt 128.0108 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity