missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenethyl alcohol, 99%, Thermo Scientific Chemicals

Catalog Number p-76448
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1074-16-4
C8H9BrO
201.06
MFCD00093566
ADLOWZRDUHSVRU-UHFFFAOYSA-N
2-2-bromophenyl ethanol, 2-bromophenethyl alcohol, 2-bromophenethylalcohol, 2-2-bromophenyl ethan-1-ol, benzeneethanol, 2-bromo, o-bromophenethyl alcohol, 2-2-bromophenyl ethyl alcohol, 2-bromobenzeneethanol, 2-bromophenylethanol, pubchem16178
2734089
2-(2-bromophenyl)ethanol
C1=CC=C(C(=C1)CCO)Br
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1074-16-4
C8H9BrO
201.06
MFCD00093566
ADLOWZRDUHSVRU-UHFFFAOYSA-N
2-2-bromophenyl ethanol, 2-bromophenethyl alcohol, 2-bromophenethylalcohol, 2-2-bromophenyl ethan-1-ol, benzeneethanol, 2-bromo, o-bromophenethyl alcohol, 2-2-bromophenyl ethyl alcohol, 2-bromobenzeneethanol, 2-bromophenylethanol, pubchem16178
2734089
2-(2-bromophenyl)ethanol
C1=CC=C(C(=C1)CCO)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 1.4830g/mL
Boiling Point 97.0°C (0.7 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4CH2CH2OH
Refractive Index 1.5755 to 1.5775
Quantity 1 g
Specific Gravity 1.483
Formula Weight 201.06
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Bromophenethyl alcohol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromophenethylalcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'