Få mere at vide
2-Bromoisobutyric-d6 Acid, TRC
Artikelnummer. 30334730
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30334730 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30334730 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68464725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Bromoisobutyric-d6 Acid
CAS 1219795-23-9
Molekylær formel C4 D6 H Br O2
Mængde 25 mg
Synonym 2-Bromo-2-methyl-propanoic-d6 Acid, α-Bromo-isobutyric-d6 Acid, 2-Bromo-2-methyl-propionic-d6 Acid, 2-Bromo-2-methylpropanoic-d6 Acid, 2-Bromo-2-methylpropionic-d6 Acid, 2-Bromoisobutyric-d6 Acid, Bromoisobutyric-d6 Acid, Isobromobutyric-d6 Acid, NSC 41213-d6, α-Bromo-α-methylpropionic-d6 Acid, α-Bromoisobutyric-d6 Acid
InChI formel InChI=1S/C4H7BrO2/c1-4(2,5)3(6)7/h1-2H3,(H,6,7)/i1D3,2D3
SMIL [2H]C([2H])([2H])C(Br)(C(=O)O)C([2H])([2H])[2H]
IUPAC navn 2-bromo-3,3,3-trideuterio-2-(trideuteriomethyl)propanoic acid
Molekylvægt (g/mol) 173.04
Formel vægt 172.0006