missing translation for 'onlineSavingsMsg'
Learn More

2-Bromofluorobenzene, 99%, Thermo Scientific Chemicals

Catalog Number p-74551
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
1072-85-1
C6H4BrF
175
MFCD00000282
IPWBFGUBXWMIPR-UHFFFAOYSA-N
2-bromofluorobenzene, o-bromofluorobenzene, 2-fluorobromobenzene, o-fluorobromobenzene, 2-bromfluorbenzen, benzene, 1-bromo-2-fluoro, 1-fluoro-2-bromobenzene, benzene, 1-fluoro-2-bromo, ofbb, benzene, bromofluoro
61259
1-bromo-2-fluorobenzene
C1=CC=C(C(=C1)F)Br
This item is not returnable. View return policy
1072-85-1
C6H4BrF
175
MFCD00000282
IPWBFGUBXWMIPR-UHFFFAOYSA-N
2-bromofluorobenzene, o-bromofluorobenzene, 2-fluorobromobenzene, o-fluorobromobenzene, 2-bromfluorbenzen, benzene, 1-bromo-2-fluoro, 1-fluoro-2-bromobenzene, benzene, 1-fluoro-2-bromo, ofbb, benzene, bromofluoro
61259
1-bromo-2-fluorobenzene
C1=CC=C(C(=C1)F)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.6000g/mL
Boiling Point 155.0°C to 157.0°C
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula BrC6H4F
Refractive Index 1.5327 to 1.5347
Quantity 10 mL
Fieser 01,77
Specific Gravity 1.6
Solubility Information Solubility in water: insoluble in water
Formula Weight 175
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Bromofluorobenzene, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'