missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals

Catalog Number p-74073
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
2576-47-8, 107-09-5
C2H6BrN·HBr
204.9
MFCD00012886
WJAXXWSZNSFVNG-UHFFFAOYSA-N
2-bromoethylamine hydrobromide, 2-bromoethanamine hydrobromide, 2-aminoethyl bromide hydrobromide, 2-bromoethylamine hbr, 2-bromoethylammonium bromide, ethanamine, 2-bromo-, hydrobromide, 2-bromoethan-1-amine hydrobromide, bromoethylamine hydrobromide, aminoethyl bromide hbr, ethylamine, hydrobromide
2774217
2-bromoethanamine;hydrobromide
C(CBr)N.Br
This item is not returnable. View return policy
2576-47-8, 107-09-5
C2H6BrN·HBr
204.9
MFCD00012886
WJAXXWSZNSFVNG-UHFFFAOYSA-N
2-bromoethylamine hydrobromide, 2-bromoethanamine hydrobromide, 2-aminoethyl bromide hydrobromide, 2-bromoethylamine hbr, 2-bromoethylammonium bromide, ethanamine, 2-bromo-, hydrobromide, 2-bromoethan-1-amine hydrobromide, bromoethylamine hydrobromide, aminoethyl bromide hbr, ethylamine, hydrobromide
2774217
2-bromoethanamine;hydrobromide
C(CBr)N.Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 172.0°C to 176.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Plastic bottle
Linear Formula BrCH2CH2NH2·HBr
Quantity 25 g
Beilstein 04,134
Solubility Information Solubility in water: >500g/L (20°C)
Formula Weight 204.9
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Bromoethylamine hydrobromide, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'