missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethyl methyl ether, 96%, stab. with sodium carbonate, Thermo Scientific Chemicals

Catalog Number p-7022542
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
6482-24-2
C3H7BrO
138.992
MFCD00000236
YZUPZGFPHUVJKC-UHFFFAOYSA-N
2-bromoethyl methyl ether, 2-methoxyethyl bromide, ethane, 1-bromo-2-methoxy, ether, 2-bromoethyl methyl, methoxyethyl bromide, 2-bromoethylmethylether, bromoethyl methyl ether, 2-bromoethylmethyl ether, 2-bromoethylmethyl-ether, 2-bromo-1-methoxyethane
80972
1-bromo-2-methoxyethane
COCCBr
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
6482-24-2
C3H7BrO
138.992
MFCD00000236
YZUPZGFPHUVJKC-UHFFFAOYSA-N
2-bromoethyl methyl ether, 2-methoxyethyl bromide, ethane, 1-bromo-2-methoxy, ether, 2-bromoethyl methyl, methoxyethyl bromide, 2-bromoethylmethylether, bromoethyl methyl ether, 2-bromoethylmethyl ether, 2-bromoethylmethyl-ether, 2-bromo-1-methoxyethane
80972
1-bromo-2-methoxyethane
COCCBr

2-Bromoethyl methyl ether has been used as starting reagent in the synthesis of 1-bromo-4-(2-methoxyethoxy)benzene1 and 1-bromo-4-[(2-methoxyethoxy)methyl]benzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoethyl methyl ether has been used as starting reagent in the synthesis of 1-bromo-4-(2-methoxyethoxy)benzene1 and 1-bromo-4-[(2-methoxyethoxy)methyl]benzene.

Solubility
Soluble in water.14,4 g/L at 25°C

Notes
Protect from heat. Incompatible with heat, strong bases and oxidizing agents. Store at 2-8°C.
TRUSTED_SUSTAINABILITY
Density 1.478
Boiling Point 109°C to 110°C
Flash Point 28°C (82°F)
Odor Recognizable
Refractive Index 1.448
Quantity 5 g
UN Number UN3271
Solubility Information Soluble in water.14,4g/L at 25°C
Formula Weight 139
Percent Purity 96%
Chemical Name or Material 2-Bromoethyl methyl ether, stabilized with sodium carbonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'