Få mere at vide
2-Bromoethyl-d4-amine HBr, CDN
Artikelnummer. 30325257
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30325257 0.1 g 0.1g
30322390 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325257 Leverandør CDN Leverandørnr. CDND66710.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Bromoethylamine-D4 Hydrobromide
CAS 918633-70-2
Molekylær formel C2 D4 H2 Br N . Br H
Mængde 0.1 g
Synonym Ethan-1,1,2,2-d4-amine, 2-bromo-, hydrobromide (1:1) (9 CI), 2-Bromoethyl-d4-amine hydrobromide, 2-Bromoethan-1,1,2,2-d4-amine hydrobromide, 2-Bromoethylamine-D4 hydrobromide
InChI formel InChI=1 S/C2H6BrN.BrH/c3-1-2-4;/h1-2,4H2;1 H/i1D2,2D2;
SMIL Br.[2 H]C([2 H])(N)C([2 H])([2 H])Br
IUPAC navn 2-bromo-1,1,2,2-tetradeuterioethanamine;hydrobromide
Molekylvægt (g/mol) 208.9163
Formel vægt 206.9196 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity