Learn More
2-Bromoethyl acrylate, 94%, stab. with 900-1500ppm 4-methoxyphenol, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
2-Bromoethyl acrylate is utilized in curable and reactive polymers due to the presence of reactive bromine. It is also used in the synthesis of other monomers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromoethyl acrylate is utilized in curable and reactive polymers due to the presence of reactive bromine. It is also used in the synthesis of other monomers.
Solubility
Miscible with water.
Notes
Light and heat sensitive Incompatible with strong oxidizing agents, polymerizing agents and peroxides.
2-Bromoethyl acrylate is utilized in curable and reactive polymers due to the presence of reactive bromine. It is also used in the synthesis of other monomers.
Solubility
Miscible with water.
Notes
Light and heat sensitive Incompatible with strong oxidizing agents, polymerizing agents and peroxides.
Specifications
Boiling Point | 52°C to 53°C (5 mmHg) |
Quantity | 1 g |
Beilstein | 1749713 |
Synonym | 2-bromoethyl acrylate, acrylic acid, 2-bromoethyl ester, 2-propenoic acid, 2-bromoethyl ester, ccris 4753, acrylic acid 2-bromoethyl ester, 2-bromoethylacrylate, 2-bromoethyl=acrylate, acmc-20ao1p, acrylic acid 2-bromo-ethyl ester, 2-propenoic acid,2-bromoethyl ester |
Solubility Information | Miscible with water. |
Formula Weight | 179.01 |
Percent Purity | 94% |
Chemical Name or Material | 2-Bromoethyl acrylate, stabilized with 0.1% 4-methoxyphenol |