Få mere at vide
2-Bromoethyl-4-nitrophenyl Ether-d4, TRC
Artikelnummer. 30339716
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30339716 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30339716 Leverandør Toronto Research Chemicals Leverandørnr. TRCB684262250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Bromoethyl-4-nitrophenyl Ether-d4
CAS 1190017-10-7
Molekylær formel C8H4D4BrNO3
Mængde 250 mg
Synonym 1-(2-Bromoethoxy-d4)-4-nitro-benzene, beta-Bromo-4-nitrophenetole-d4, 1-(2-Bromoethoxy-d4)-4-nitrobenzene, 2-(4-Nitrophenoxy)-1-bromoethane-d4, NSC 37983-d4, p-Nitrophenyl beta-Bromoethyl-d4 Ether
InChI formel InChI=1S/C8H8BrNO3/c9-5-6-13-8-3-1-7(2-4-8)10(11)12/h1-4H,5-6H2/i5D2,6D2
SMIL [2H]C([2H])(Br)C([2H])([2H])Oc1ccc(cc1)[N+](=O)[O-]
IUPAC navn 1-(2-bromo-1,1,2,2-tetradeuterioethoxy)-4-nitrobenzene
Molekylvægt (g/mol) 250.08
Formel vægt 248.9939