Få mere at vide
2-Bromochlorobenzene-d4, CDN
Artikelnummer. 30323135
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30323135 1 g -
30327782 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323135 Leverandør CDN Leverandørnr. CDND64441G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Bromo-6-chlorobenzene-1,2,3,4-d4
CAS 1219795-51-3
Molekylær formel C6 D4 Br Cl
Mængde 1 g
Synonym 2-Bromochlorobenzene-d4, Benzene-1,2,3,4-d4, 5-bromo-6-chloro- (ACI), 5-Bromo-6-chlorobenzene-1,2,3,4-d4 (ACI)
InChI formel InChI=1 S/C6H4BrCl/c7-5-3-1-2-4-6(5)8/h1-4 H/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c([2 H])c(Br)c(Cl)c1[2 H]
IUPAC navn 1-bromo-2-chloro-3,4,5,6-tetradeuteriobenzene
Molekylvægt (g/mol) 195.48
Formel vægt 193.9436 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity