Få mere at vide
(±)-2-Bromobutane-d9, CDN
Artikelnummer. 30315547
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30315547 0.25 g -
30314849 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315547 Leverandør CDN Leverandørnr. CDND59060.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Bromobutane-d9
CAS 202392-72-1
Molekylær formel C4D9Br
Mængde 0.25 g
Synonym (±)-2-Bromobutane-d9, (±)-sec-Butyl bromide-d9, 2-Bromobutane-d9, 2-Butyl bromide-d9, Methylethylbromomethane-d9, NSC 8417-d9, sec-Butyl bromide-d9
InChI formel InChI=1 S/C4H9Br/c1-3-4(2)5/h4H,3H2,1-2H3/i1D3,2D3,3D2,4 D
SMIL BrC(C([2 H])([2 H])C([2 H])([2 H])[2 H])([2 H])C([2 H])([2 H])[2 H]
IUPAC navn 2-bromo-1,1,1,2,3,3,4,4,4-nonadeuteriobutane
Molekylvægt (g/mol) 146.07
Formel vægt 145.05 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity