missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Bromo-N-[N-[(1,1-dimethylethoxy)carbonyl]-L-alanyl]-N-methyl-D-tryptophan Methyl Ester, TRC

Artikelnummer. 30337325
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30337325 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30337325 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6819152.5MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 2-Bromo-N-[N-[(1,1-dimethylethoxy)carbonyl]-L-alanyl]-N-methyl-D-tryptophan Methyl Ester
CAS 134781-89-8
Molekylær formel C21 H28 Br N3 O5
Mængde 2.5 mg
InChI formel InChI=1S/C21H28BrN3O5/c1-12(23-20(28)30-21(2,3)4)18(26)25(5)16(19(27)29-6)11-14-13-9-7-8-10-15(13)24-17(14)22/h7-10,12,16,24H,11H2,1-6H3,(H,23,28)/t12-,16?/m0/s1
SMIL COC(=O)C(Cc1c(Br)[nH]c2ccccc12)N(C)C(=O)[C@H](C)NC(=O)OC(C)(C)C
IUPAC navn methyl 3-(2-bromo-1H-indol-3-yl)-2-[methyl-[(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoyl]amino]propanoate
Molekylvægt (g/mol) 482.368
Formel vægt 481.121
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.