missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 2-Bromo-N-(1-naphthyl)benzenesulfonamide, 97%
Brand: Alfa Aesar™ H60769.03
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
851215-65-1 | |
362.241 | |
LSOKWJIMCSVKBL-UHFFFAOYSA-N | |
2465063 | |
C1=CC=C2C(=C1)C=CC=C2NS(=O)(=O)C3=CC=CC=C3Br |
C16H12BrNO2S | |
MFCD05617821 | |
2-bromo-n-naphthalen-1-yl benzenesulfonamide, 2-bromo-n-1-naphthyl benzenesulfonamide, 2-bromo-n-naphthalen-1-yl benzene-1-sulfonamide | |
2-bromo-N-naphthalen-1-ylbenzenesulfonamide |
Specifications
2-Bromo-N-(1-naphthyl)benzenesulfonamide | |
851215-65-1 | |
MFCD05617821 | |
LSOKWJIMCSVKBL-UHFFFAOYSA-N | |
2-bromo-N-naphthalen-1-ylbenzenesulfonamide | |
2465063 | |
97% |
1g | |
C16H12BrNO2S | |
2-bromo-n-naphthalen-1-yl benzenesulfonamide, 2-bromo-n-1-naphthyl benzenesulfonamide, 2-bromo-n-naphthalen-1-yl benzene-1-sulfonamide | |
C1=CC=C2C(=C1)C=CC=C2NS(=O)(=O)C3=CC=CC=C3Br | |
362.241 | |
362.24 |