missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Bromo-6-methyl-3-nitropyridine, 98%

Brand:  Alfa Aesar™ H64982.06

Additional Details : CAS Number : 374633-31-5 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15482419

  • 1364.81 DKK / 5g


Estimated Shipment
26-10-2020
Add to basket

Description

Description

Applications
It is used as pharmaceutical intermediate.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

374633-31-5
217.022
IHHLJCDGOSZFTB-UHFFFAOYSA-N
24728013
CC1=NC(=C(C=C1)[N+](=O)[O-])Br
C6H5BrN2O2
MFCD03095212
2-bromo-3-nitro-6-methylpyridine, 2-bromo-3-nitro-6-picoline, 2-bromo-6-methyl-3-nitro-pyridine, 2-bromo-3-nitro-6-methyl pyridine, pubchem5954, acmc-209it3, ksc496c9p, 6-bromo-5-nitro-2-picoline, pyridine, 2-bromo-6-methyl-3-nitro, 2-bromo-6-methyl-3-nitropyridinemethyl 6-bromomethyl nicotinate
2-bromo-6-methyl-3-nitropyridine

Specifications

2-Bromo-6-methyl-3-nitropyridine
5g
C6H5BrN2O2
138200
IHHLJCDGOSZFTB-UHFFFAOYSA-N
2-bromo-6-methyl-3-nitropyridine
24728013
Solid
White
374633-31-5
MFCD03095212
2-bromo-3-nitro-6-methylpyridine, 2-bromo-3-nitro-6-picoline, 2-bromo-6-methyl-3-nitro-pyridine, 2-bromo-3-nitro-6-methyl pyridine, pubchem5954, acmc-209it3, ksc496c9p, 6-bromo-5-nitro-2-picoline, pyridine, 2-bromo-6-methyl-3-nitro, 2-bromo-6-methyl-3-nitropyridinemethyl 6-bromomethyl nicotinate
CC1=NC(=C(C=C1)[N+](=O)[O-])Br
217.022
217.02
98%
Videos
SDS Documents