missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Bromo-6-fluoro-4-methylpyridine, 98%

Brand:  Alfa Aesar™ H64449.06

Additional Details : CAS Number : 180608-37-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15401669

  • 5886.77 DKK / 5g


In Stock 1
Add to basket

Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

2-Bromo-6-fluoro-4-methylpyridine
MFCD09839269
2-Bromo-6-fluoro-4-picoline
180608-37-1
13574124

Specifications

2-Bromo-6-fluoro-4-methylpyridine
180608-37-1
Liquid
5g
MFCD09839269
2-Bromo-6-fluoro-4-picoline
CC1=CC(=NC(=C1)Br)F
190.015
190.02
Colorless to pale yellow
98%
C6H5BrFN
13574124
CIDWWTCSJWQJQR-UHFFFAOYSA-N
2-bromo-6-fluoro-4-methylpyridine
29919366
Videos
SDS