missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Bromo-5-chlorobenzaldehyde, 97%

Brand:  Alfa Aesar™ H64330.22

Additional Details : CAS Number : 174265-12-4 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 15471489

  • 2863.86 DKK / 100g


Estimated Shipment date
from Supplier 23-06-2020
Add to basket

Specifications

Chemical Identifiers

174265-12-4
219.462
IIISHLMCTDMUHH-UHFFFAOYSA-N
15391279
C1=CC(=C(C=C1Cl)C=O)Br
C7H4BrClO
MFCD00462870
5-chloro-2-bromobenzaldehyde, benzaldehyde, 2-bromo-5-chloro, pubchem19849, acmc-1bwmm, ksc174i5n, 2-bromo-5-chlorobenzaldehycle, iiishlmctdmuhh-uhfffaoysa
2-bromo-5-chlorobenzaldehyde

Specifications

2-Bromo-5-chlorobenzaldehyde
64°C to 68°C (literature)
174265-12-4
MFCD00462870
5-chloro-2-bromobenzaldehyde, benzaldehyde, 2-bromo-5-chloro, pubchem19849, acmc-1bwmm, ksc174i5n, 2-bromo-5-chlorobenzaldehycle, iiishlmctdmuhh-uhfffaoysa
C1=CC(=C(C=C1Cl)C=O)Br
219.462
219.46
97%
Yellow
100g
C7H4BrClO
10423891
IIISHLMCTDMUHH-UHFFFAOYSA-N
2-bromo-5-chlorobenzaldehyde
15391279
Crystalline
Videos
SDS Documents