missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4-(trifluoromethyl)phenylacetonitrile, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
2-Bromo-4-(trifluoromethyl)phenylacetonitrile is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-4-(trifluoromethyl)phenylacetonitrile is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
2-Bromo-4-(trifluoromethyl)phenylacetonitrile is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications
Density | 1.626 |
Boiling Point | 265°C to 266°C |
Refractive Index | 1.502 |
Quantity | 1 g |
Synonym | 2-bromo-4-trifluoromethyl phenylacetonitrile, 2-2-bromo-4-trifluoromethyl phenyl acetonitrile, 2-2-bromo-4-trifluoromethyl phenyl ethanenitrile, pubchem4248, 2-bromo-4-trifluoromethyl phenyl acetonitrile, benzeneacetonitrile,2-bromo-4-trifluoromethyl, benzeneacetonitrile, 2-bromo-4-trifluoromethyl |
Solubility Information | Slightly soluble in water. |
Formula Weight | 264.04 |
Percent Purity | 98% |
Chemical Name or Material | 2-Bromo-4-(trifluoromethyl)phenylacetonitrile |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'