Få mere at vide
2-Bromo-4’-methylacetophenone-d3, TRC
Artikelnummer. 30332765
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30332765 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30332765 Leverandør Toronto Research Chemicals Leverandørnr. TRCB684952100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Bromo-4'-methylacetophenone-d3
CAS 959605-55-1
Molekylær formel C9 2H3 H6 Br O
Mængde 100 mg
Synonym 4-Methylphenacyl-d3 Bromide, 2-Bromo-1-(4-methylphenyl)ethanone-d3, Bromomethyl p-Tolyl Ketone-d3, NSC 63192-d3
InChI formel InChI=1S/C9H9BrO/c1-7-2-4-8(5-3-7)9(11)6-10/h2-5H,6H2,1H3/i1D3
SMIL [2H]C([2H])([2H])c1ccc(cc1)C(=O)CBr
IUPAC navn 2-bromo-1-[4-(trideuteriomethyl)phenyl]ethanone
Molekylvægt (g/mol) 216.09
Formel vægt 215.003