missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluorobenzyl bromide, 96%, Thermo Scientific™

Catalog Number p-7054168
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
61150-57-0
C7H5Br2F
267.92
MFCD03840524
QPLUIZXBWYUFMY-UHFFFAOYSA-N
2-bromo-4-fluorobenzyl bromide, 2-bromo-4-fluorobenzylbromide, 2-bromo-1-bromomethyl-4-fluorobenzene, benzene, 2-bromo-1-bromomethyl-4-fluoro, 4-fluoro-2-bromobenzylbromide, 1-bromo-2-bromomethyl-5-fluorobenzene, pubchem4883, 2-bromo-1-bromomethyl-4-fluoro-benzene, acmc-1b995
10706977
2-bromo-1-(bromomethyl)-4-fluorobenzene
FC1=CC(Br)=C(CBr)C=C1
This item is not returnable. View return policy
61150-57-0
C7H5Br2F
267.92
MFCD03840524
QPLUIZXBWYUFMY-UHFFFAOYSA-N
2-bromo-4-fluorobenzyl bromide, 2-bromo-4-fluorobenzylbromide, 2-bromo-1-bromomethyl-4-fluorobenzene, benzene, 2-bromo-1-bromomethyl-4-fluoro, 4-fluoro-2-bromobenzylbromide, 1-bromo-2-bromomethyl-5-fluorobenzene, pubchem4883, 2-bromo-1-bromomethyl-4-fluoro-benzene, acmc-1b995
10706977
2-bromo-1-(bromomethyl)-4-fluorobenzene
FC1=CC(Br)=C(CBr)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN3261
Formula Weight 267.92
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-Bromo-4-fluorobenzyl bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'