missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4'-fluoroacetophenone, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Description
2-Bromo-4'-fluoroacetophenone is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-4′-fluoroacetophenone is used as an organic chemical synthesis intermediate.
Solubility
Soluble in methanol.
Notes
Moisture sensitive. Incompatible with oxidizing agents.
2-Bromo-4′-fluoroacetophenone is used as an organic chemical synthesis intermediate.
Solubility
Soluble in methanol.
Notes
Moisture sensitive. Incompatible with oxidizing agents.
Specifications
Melting Point | 45°C to 49°C |
Boiling Point | 150°C (12 mmHg) |
Flash Point | >110°C (230°F) |
Quantity | 100 g |
Beilstein | 637863 |
Solubility Information | Soluble in methanol. |
Formula Weight | 217.04 |
Percent Purity | 97% |
Chemical Name or Material | 2-Bromo-4'-fluoroacetophenone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'