missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,5-difluorophenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog Number 15480859
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
883502-07-6
C8H5BrF2O2
251.03
MFCD04973423
FQSLXVRCDYUESO-UHFFFAOYSA-N
2-bromo-4,5-difluorophenylacetic acid, 2-2-bromo-4,5-difluorophenyl acetic acid, 2-bromo-4,5-difluorophenyl acetic acid, 2-bromo-4,5-difluorophenylaceticacid, pubchem2702, acmc-209qsj, 2-2-bromo-4,5-difluorophenyl aceticacid, benzeneacetic acid, 2-bromo-4,5-difluoro
3732964
2-(2-bromo-4,5-difluorophenyl)acetic acid
OC(=O)CC1=CC(F)=C(F)C=C1Br
This item is not returnable. View return policy
883502-07-6
C8H5BrF2O2
251.03
MFCD04973423
FQSLXVRCDYUESO-UHFFFAOYSA-N
2-bromo-4,5-difluorophenylacetic acid, 2-2-bromo-4,5-difluorophenyl acetic acid, 2-bromo-4,5-difluorophenyl acetic acid, 2-bromo-4,5-difluorophenylaceticacid, pubchem2702, acmc-209qsj, 2-2-bromo-4,5-difluorophenyl aceticacid, benzeneacetic acid, 2-bromo-4,5-difluoro
3732964
2-(2-bromo-4,5-difluorophenyl)acetic acid
OC(=O)CC1=CC(F)=C(F)C=C1Br

2-Bromo-4,5-difluorophenylacetic acid is used as intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4,5-difluorophenylacetic acid is used as intermediate in organic syntheses.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 251.03
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-4,5-difluorophenylacetic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'