missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4'-(1-pyrrolidinyl)acetophenone, 97%, Thermo Scientific Chemicals
420.81 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
2-Bromo-4'-(1-pyrrolidinyl)acetophenone is an important raw material and used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-4′-(1-pyrrolidinyl)acetophenone is an important raw material and used as pharmaceutical intermediates.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
2-Bromo-4′-(1-pyrrolidinyl)acetophenone is an important raw material and used as pharmaceutical intermediates.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
Specifications
Melting Point | 120°C to 122°C |
Quantity | 5 g |
UN Number | UN3261 |
Beilstein | 8549816 |
Solubility Information | Insoluble in water. |
Formula Weight | 268.16 |
Percent Purity | 97% |
Chemical Name or Material | 2-Bromo-4'-(1-pyrrolidinyl)acetophenone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'