missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Bromo-4-(1,1,3,3-tetramethylbutyl)phenol, TRC

Artikelnummer. 30348251
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30348251 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30348251 Leverandør Toronto Research Chemicals Leverandørnr. TRCB688483100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 2-Bromo-4-(1,1,3,3-tetramethylbutyl)phenol
CAS 57835-35-5
Molekylær formel C14H21BrO
Mængde 100 mg
InChI formel InChI=1S/C14H21BrO/c1-13(2,3)9-14(4,5)10-6-7-12(16)11(15)8-10/h6-8,16H,9H2,1-5H3
SMIL CC(C)(C)CC(C)(C)c1ccc(O)c(Br)c1
IUPAC navn 2-bromo-4-(2,4,4-trimethylpentan-2-yl)phenol
Molekylvægt (g/mol) 285.22
Formel vægt 284.0776
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.