Få mere at vide
2-Bromo-3'-methoxyacetophenone-13CD3, TRC
Artikelnummer. 30331555
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30331555 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30331555 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68480225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Bromo-3'-methoxyacetophenone-13CD3
CAS 1329616-33-2
Molekylær formel C813CH6D3BrO2
Mængde 25 mg
Synonym 3'-Methoxyphenacyl-13CD3 Bromide, 2-Bromo-1-(3-methoxyphenyl)ethanone-13CD3, 3-Methoxyphenacyl-13CD3 Bromide, NSC 405833-13CD3
InChI formel InChI=1S/C9H9BrO2/c1-12-8-4-2-3-7(5-8)9(11)6-10/h2-5H,6H2,1H3/i1+1D3
SMIL [2H][13C]([2H])([2H])Oc1cccc(c1)C(=O)CBr
IUPAC navn 2-bromo-1-[3-(trideuterio(1^{13}C)methoxy)phenyl]ethanone
Molekylvægt (g/mol) 233.08
Formel vægt 232.0008