Få mere at vide
2-Bromo-3-benzyloxy-4-methoxybenzoic Acid, TRC
Artikelnummer. 30343332
Skift visning
Mængde:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30343332 250 mg
30339483 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30343332 Leverandør Toronto Research Chemicals Leverandørnr. TRCB680275250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Bromo-3-benzyloxy-4-methoxybenzoic Acid
CAS 135586-17-3
Molekylær formel C15H13BrO4
Mængde 250 mg
Synonym 2-Bromo-4-methoxy-3-(phenylmethoxy)-benzoic Acid
InChI formel InChI=1S/C15H13BrO4/c1-19-12-8-7-11(15(17)18)13(16)14(12)20-9-10-5-3-2-4-6-10/h2-8H,9H2,1H3,(H,17,18)
SMIL COc1ccc(C(=O)O)c(Br)c1OCc2ccccc2
IUPAC navn 2-bromo-4-methoxy-3-phenylmethoxybenzoic acid
Molekylvægt (g/mol) 337.17
Formel vægt 335.9997