missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3,6-difluorobenzaldehyde, 96%, Thermo Scientific™

Catalog Number p-7034630
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
934987-26-5
C7H3BrF2O
221.00
MFCD12922618
PQXMBFZKKQNHBA-UHFFFAOYSA-N
benzaldehyde, 2-bromo-3,6-difluoro
50998399
2-bromo-3,6-difluorobenzaldehyde
FC1=C(Br)C(C=O)=C(F)C=C1
This item is not returnable. View return policy
934987-26-5
C7H3BrF2O
221.00
MFCD12922618
PQXMBFZKKQNHBA-UHFFFAOYSA-N
benzaldehyde, 2-bromo-3,6-difluoro
50998399
2-bromo-3,6-difluorobenzaldehyde
FC1=C(Br)C(C=O)=C(F)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Bromo-3,6-difluorobenzaldehyde is used as an organic chemical synthesis intermediate.

Notes
Air sensitive. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY
Melting Point 49°C to 51°C
Quantity 250 mg
Sensitivity Air sensitive
Formula Weight 221
Percent Purity 96%
Chemical Name or Material 2-Bromo-3,6-difluorobenzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'